Lectin PHA-L From <i>Phaseolus vulgaris </i>(red kidney bean), Alexa Fluor&trade; 594 Conjugate
Invitrogen™
Lectin PHA-L From Phaseolus vulgaris (red kidney bean), Alexa Fluor™ 594 Conjugate
Lectin phytohemagglutinin-L (PHA-L) is strongly inhibited by N-acetylglucosamine β(1-2) mannopyranosyl residues. This lectin has been shown to agglutinate leukocytes andRead more
Have Questions?
Catalog NumberQuantity
L324561 mg
Catalog number L32456
Price (USD)
204.00
Each
Quantity:
1 mg
Price (USD)
204.00
Each
Lectin phytohemagglutinin-L (PHA-L) is strongly inhibited by N-acetylglucosamine β(1-2) mannopyranosyl residues. This lectin has been shown to agglutinate leukocytes and induce mitogenic activity in T-lymphocytes and has also been used as an anterograde tracer for the study of neuronal pathways.
View complete list of fluorescent dye-conjugated lectins ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeLectins
Quantity1 mg
Shipping ConditionRoom Temperature
SourceRed kidney bean
ConjugateAlexa Fluor 594
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (2)
Citations & References
Abstract
A proposal for a coordinated effort for the determination of brainwide neuroanatomical connectivity in model organisms at a mesoscopic scale.
Authors:Bohland JW, Wu C, Barbas H, Bokil H, Bota M, Breiter HC, Cline HT, Doyle JC, Freed PJ, Greenspan RJ, Haber SN, Hawrylycz M, Herrera DG, Hilgetag CC, Huang ZJ, Jones A, Jones EG, Karten HJ, Kleinfeld D, Kötter R, Lester HA, Lin JM, Mensh BD, Mikula S, Panksepp J, Price JL, Safdieh J, Saper CB, Schiff ND, Schmahmann JD, Stillman BW, Svoboda K, Swanson LW, Toga AW, Van Essen DC, Watson JD, Mitra PP,
Journal:PLoS Comput Biol
PubMed ID:19325892
In this era of complete genomes, our knowledge of neuroanatomical circuitry remains surprisingly sparse. Such knowledge is critical, however, for both basic and clinical research into brain function. Here we advocate for a concerted effort to fill this gap, through systematic, experimental mapping of neural circuits at a mesoscopic scale ... More
Social motility of biofilm-like microcolonies in a gliding bacterium.
Authors:
Journal:Nat Commun
PubMed ID:34588437