Lectin PNA From Arachis hypogaea (peanut), Alexa Fluor™ 568 Conjugate
Lectin PNA is specific for terminal β-galactose. It will agglutinate human erythrocytes, but only after neuraminidase treatment.View complete list ofRead more
Have Questions?
Catalog Number
Quantity
L32458
1 mg
Catalog number L32458
Price (TWD)
6,600.00
Online offer
Ends: 31-Dec-2026
9,420.00
Save 2,820.00 (30%)
Each
Quantity:
1 mg
Price (TWD)
6,600.00
Online offer
Ends: 31-Dec-2026
9,420.00
Save 2,820.00 (30%)
Each
Lectin PNA is specific for terminal β-galactose. It will agglutinate human erythrocytes, but only after neuraminidase treatment.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeLectins
Quantity1 mg
Shipping ConditionRoom Temperature
SourcePeanut
ConjugateAlexa Fluor 568
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (11)
Citations & References
Abstract
Identification of a novel signal in the cytoplasmic tail of the Na+:HCO3- cotransporter NBC1 that mediates basolateral targeting.
Authors:Li HC, Li EY, Neumeier L, Conforti L, Soleimani M,
Journal:Am J Physiol Renal Physiol
PubMed ID:17182531
'The Na(+):HCO(3)(-) cotransporter NBC1 (SLC4A4, variant A, kidney specific) is located exclusively on the basolateral membrane of epithelial cells, implying that this molecule has acquired specific signals for targeting to the basolateral membrane. A motif with the sequence QQPFLS (positions 1010-1015) in the cytoplasmic tail of NBC1 was recently demonstrated ... More
Merging organoid and organ-on-a-chip technology to generate complex multi-layer tissue models in a human retina-on-a-chip platform.
Authors:
Journal:Elife
PubMed ID:31451149
Chd5 orchestrates chromatin remodelling during sperm development.
Authors:
Journal:Nat Commun
PubMed ID:24818823
3D in situ imaging of the female reproductive tract reveals molecular signatures of fertilizing spermatozoa in mice.
Authors:
Journal:Elife
PubMed ID:33078708
Absence of CFAP69 Causes Male Infertility due to Multiple Morphological Abnormalities of the Flagella in Human and Mouse.