Lectin PNA From <i>Arachis hypogaea </i>(peanut), Alexa Fluor&trade; 594 Conjugate
Invitrogen™
Lectin PNA From Arachis hypogaea (peanut), Alexa Fluor™ 594 Conjugate
Lectin PNA is specific for terminal β-galactose. It will agglutinate human erythrocytes, but only after neuraminidase treatment.View complete list ofRead more
Have Questions?
Catalog NumberQuantity
L324591 mg
Catalog number L32459
Price (USD)
182.04
Special offer
Online exclusive
Ends: 31-Dec-2026
222.00
Save 39.96
Each
Quantity:
1 mg
Price (USD)
182.04
Special offer
Online exclusive
Ends: 31-Dec-2026
222.00
Save 39.96
Each
Lectin PNA is specific for terminal β-galactose. It will agglutinate human erythrocytes, but only after neuraminidase treatment.
View complete list of fluorescent dye-conjugated lectins ›
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeAlexa Fluor Dyes
Product LineAlexa Fluor
Protein SubtypeLectins
Quantity1 mg
Shipping ConditionRoom Temperature
SourcePeanut
ConjugateAlexa Fluor 594
FormLyophilized
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (10)
Citations & References
Abstract
Localization of sphingosine kinase-1 in mouse sperm acrosomes.
Authors:Matsumoto K, Banno Y, Murate T, Akao Y, Nozawa Y
Journal:J Histochem Cytochem
PubMed ID:15684337
Sphingosine kinase (SPHK) catalyzes sphingosine phosphorylation to form a bioactive lipid mediator, sphingosine-1-phosphate (S1P). In the current study, we report the presence of SPHK-1 in mouse spermatozoa. SPHK-1 was localized to the acrosomes of spermatozoa, and its expression was proven by RT-PCR and Western blot analysis. SPHK activity of mouse ... More
Substantial restoration of night vision in adult mice with congenital stationary night blindness.
Authors:
Journal:Mol Ther Methods Clin Dev
PubMed ID:34401402
Shared developmental programme strongly constrains beak shape diversity in songbirds.
Authors:
Journal:Nat Commun
PubMed ID:24739280
Therapeutic Regeneration of Lymphatic and Immune Cell Functions upon Lympho-organoid Transplantation.
Authors:
Journal:Stem Cell Reports
PubMed ID:31155505
Natural history of cone disease in the murine model of Leber congenital amaurosis due to CEP290 mutation: determining the timing and expectation of therapy.
Authors:
Journal:PLoS One
PubMed ID:24671090