Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 594
Invitrogen™

Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 594

Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 594, is a bright-red LEL-fluorophore conjugate. The tomato lectin LEL is widely used자세히 알아보기
Have Questions?
카탈로그 번호수량
L324711 mg
카탈로그 번호 L32471
제품 가격(KRW)
295,000
온라인 행사
Ends: 31-Dec-2026
337,000
할인액 42,000 (12%)
Each
수량:
1 mg
제품 가격(KRW)
295,000
온라인 행사
Ends: 31-Dec-2026
337,000
할인액 42,000 (12%)
Each
Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 594, is a bright-red LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-DyLight 594 can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.

• Very bright DyLight 594 conjugate detected with traditional Texas Red filter set, Ex/Em: 592/617 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
• Inhibiting/eluting sugar: chitin hydrolysate
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 71 KDa.
• Immunofluorescence working concentration: 5–20 μg/mL

LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.

For Research Use Only. Not for use in diagnostic procedures.
사양
라벨 유형DyLight Fluors
제품라인DyLight
단백질 아형Lectins
수량1 mg
배송 조건Approved for shipment at Room Temperature or on Wet Ice
소스Tomato
콘주게이트DyLight 594
형태Solution
Unit SizeEach
구성 및 보관
When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after receipt.