Dolichos Biflorus (Horse Gram) Agglutinin (DBA), fluorescein (FITC)
Invitrogen™

Dolichos Biflorus (Horse Gram) Agglutinin (DBA), fluorescein (FITC)

Dolichos Biflorus (Horse Gram) Agglutinin (DBA), fluorescein (FITC), is a bright-green DBA-fluorophor conjugate. DBA has been used to establish secretorRead more
Have Questions?
Catalog NumberQuantity
L324742 mg
Catalog number L32474
Price (USD)
203.30
Special offer
Online exclusive
Ends: 27-Jun-2026
214.00
Save 10.70
Each
Quantity:
2 mg
Price (USD)
203.30
Special offer
Online exclusive
Ends: 27-Jun-2026
214.00
Save 10.70
Each
Dolichos Biflorus (Horse Gram) Agglutinin (DBA), fluorescein (FITC), is a bright-green DBA-fluorophor conjugate. DBA has been used to establish secretor status in blood-group-A individuals by hemagglutination inhibition techniques and for blood typing. This lectin has also been used as a general marker of developing renal collecting ducts. It has also been used as a marker of cell surface change in murine embryonic stem cells during early differentiation.

View complete list of fluorescent dye-conjugated lectins ›

• Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: N-acetylgalactosamine
• Inhibiting/eluting sugar: 200 mM N-acetylgalactosamine
• Blood group specificity: A1 >>A2
• Unconjugated lectin molecular weight: 111 KDa
• Immunofluorescence working concentration: 5–20 μg/mL

DBA-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Protein FamilyLectins
Quantity2 mg
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
SourceHorse gram
ConjugateFITC
FormLyophilized
Unit SizeEach
Contents & Storage
When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.