Lens Culinaris (Edible Lentil) Agglutinin (LCA), fluorescein (FITC)
Invitrogen™

Lens Culinaris (Edible Lentil) Agglutinin (LCA), fluorescein (FITC)

Lens Culinaris Agglutinin (LCA), fluorescein ( FITC), is a bright-green LCA-fluorophor conjugate that is sourced from a common edible lentil.자세히 알아보기
Have Questions?
카탈로그 번호수량
L324755 mg
카탈로그 번호 L32475
제품 가격(KRW)
214,000
온라인 행사
Ends: 30-Sep-2026
244,000
할인액 30,000 (12%)
Each
수량:
5 mg
제품 가격(KRW)
214,000
온라인 행사
Ends: 30-Sep-2026
244,000
할인액 30,000 (12%)
Each
Lens Culinaris Agglutinin (LCA), fluorescein ( FITC), is a bright-green LCA-fluorophor conjugate that is sourced from a common edible lentil. LCA specificity is similar to concanavalin A, but narrower, resulting in differences in patterns of tissue staining. Wheat germ agglutinin, concanavalin A, and LCA are known to block inhibitory effects of nerve growth factor in cell-free systems.

• Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: mannose, glucose
• Inhibiting/eluting sugar: mixture of 200 mM α-methylmannoside/200 mM α-methylglucoside
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 50 KDa
• Immunofluorescence working concentration: 5–20 μg/mL

LCA-FITC is provided at a 5 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+.

If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.

For Research Use Only. Not for use in diagnostic procedures.
사양
단백질군Lectins
수량5 mg
배송 조건Approved for shipment at Room Temperature or on Wet Ice
소스Lentil
콘주게이트FITC
형태Solution
Unit SizeEach
구성 및 보관
When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after receipt.