Oxidized Low-density Lipoprotein (OxLDL) and DiI-OxLDL
Invitrogen™

Oxidized Low-density Lipoprotein (OxLDL) and DiI-OxLDL

Study lipid transport and atherosclerosis with oxidized LDL (OxLDL) and the fluorescently labeled DiI-OxLDL. Oxidized forms of human low-density lipoprotein (LDL) are used to examine scavenger receptor-mediated endocytosis of OxLDL by macrophages and endothelial cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugate
L34357Ox-LDL
L34358Dil-OxLDL
Catalog number L34357
Price (USD)
293.65
Online exclusive
309.00
Save 15.35
Each
Conjugate:
Ox-LDL
Price (USD)
293.65
Online exclusive
309.00
Save 15.35
Each
Oxidized LDL (OxLDL) and the fluorescently labeled DiI-OxLDL are oxidized forms of the human low-density lipoprotein (LDL). LDL is an important lipid-protein complex that is involved in lipid transport and contributes to atherosclerosis. These unlabeled and fluorescent OxLDL probes are used to study scavenger receptor-mediated endocytosis by macrophages and endothelial cells, and the formation of macrophage-derived foam cells that occurs in early atherogenesis.

Human low-density lipoprotein (LDL) is one of the key lipid-protein complexes in blood and is a crucial component of metabolism responsible for the transport of lipids throughout the body. LDL is composed of cholesterol, cholesterol esters, triglycerides, phospholipids, and a single Apo B-100 protein. LDLs deliver fatty acids and cholesterol to peripheral and liver cells through receptor-mediated endocytosis. High levels of LDL cholesterol are considered a risk factor for atherosclerosis and cardiovascular disease, while high levels of high-density lipoproteins (HDLs) may reduce the risk.

Oxidation of LDL is a natural process within the body caused by free radicals, however excess oxidized LDL can be harmful. Alterations in lipid metabolism such as oxidation of LDL can lead to inflammation in arteries and atherosclerosis. Atherosclerosis is a type of arteriosclerosis in which arterial wall thickening is due to the buildup of fatty plaques, composed of cholesterol and other lipids, that can inhibit and block blood flow in vessels. Endothelial cells and macrophages are recruited to remove oxidized LDL (OxLDL), which initiates inflammatory and immunogenic responses. Unlabeled and fluorescent OxLDL probes are important tools in the study of scavenger receptor-mediated endocytosis by macrophages and endothelial cells, as well as the formation of macrophage-derived foam cells, a hallmark of early atherogenesis.

The OxLDL probe is generated by oxidizing the surface lipids of native unmodified LDL by incubating with copper sulfate. Oxidation is monitored during the incubation by measuring the optical density at 234 and terminated at approximately the halfway point during the lipid peroxidation phase to a level of approximately 25–35 nmol/mg protein in TBAR (thiobarbituric acid–reactive) values. This controlled oxidation procedure is used to ensure that lipids on the surface of the LDL are oxidized, with limited oxidation of the surface apolipoprotein, allowing the OxLDL to induce a physiologically relevant inflammatory response from cells.

Features of OxLDL:
• Oxidized by copper-mediated process
• Each lot tested for optimal level of oxidation and functionally tested with Bovine Pulmonary Artery Epithelium (BPAE) cells for recognition by scavenger receptors
• Can be used as a negative control to block cell surface receptors or to stimulate inflammatory/immunogenic responses and in applications such as ELISA and Western blot

Oxidized LDL is also complexed with the red-orange fluorescent DiI dye (DiI-OxLDL) to enable fluorescence detection of oxidized LDL uptake. DiI is a highly fluorescent lipophilic dye that diffuses into the hydrophobic portion of the LDL complex and does not affect the LDL-specific binding of the apolipoprotein. The DiI-OxLDL degree of labeling has been optimized for superior sensitivity in various fluorescence applications such as fluorescence microscopy, flow cytometry, cell sorting, high-throughput analysis, and high-content analysis.

Features of DiI-OxLDL:
• Detectable with RFP/TRITC filter set (absorption peak: ∼554 nm, emission peak: ∼571 nm)
• Fixable and multiplexable with green, deep red, violet, and blue emission dyes, such as GFP, DAPI, Hoechst, HCS LipidTOX, and LysoTracker Deep Red
• Each lot functionally tested with Bovine Pulmonary Artery Epithelium (BPAE) cells for recognition by scavenger receptors
• Optimized oxidization and degree of labeling for superior sensitivity

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateOx-LDL
FormSolution
Protein FamilyLDL
Quantity200 μL
Shipping ConditionWet Ice
SpeciesHuman
For Use With (Application)Cell Viability, Cell Proliferation and Function
Product LineMolecular Probes
Unit SizeEach
Contents & Storage
1 x 200 μL OxLDL, 2.5 mg/mL
Store in refrigerator (2°C to 8°C) and protect from light.

Frequently asked questions (FAQs)

If the Low Density Lipoprotein (LDL) from Human Plasma was accidentally frozen, can I still use it?

We recommend storing Low Density Lipoprotein (LDL) from Human Plasma at 2-8 degrees C. We do not recommend using frozen product.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use Low Density Lipoprotein (LDL) from Human Plasma on other species?

We have not tested this.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (5)

Citations & References
Abstract
Stroke induces disease-specific myeloid cells in the brain parenchyma and pia.
Authors:Minnerup J
Journal:Nature communications
PubMed ID:35177618
A neutralizing antibody against DKK1 does not reduce plaque formation in classical murine models of atherosclerosis: Is the therapeutic potential lost in translation?
Authors:Nyberg M
Journal:Atherosclerosis
PubMed ID:33129080
GRP78/BiP alleviates oxLDL-induced hepatotoxicity in familial hypercholesterolemia caused by missense variants of LDLR in a HepG2 cellular model.
Authors:Ali BR
Journal:Lipids in health and disease
PubMed ID:37248472
Measurement of lipid droplets in peripheral immune cells shows an immunomodulatory effect on monocyte polarization in experimental dyslipidaemia.
Authors:Lin CF
Journal:Biochemical and biophysical research communications
PubMed ID:36773342
Chronic mitochondria antioxidant treatment in older adults alters the circulating milieu to improve endothelial cell function and mitochondrial oxidative stress.
Authors:Rossman MJ
Journal:American journal of physiology. Heart and circulatory physiology
PubMed ID:37326998