Low Density Lipoprotein from Human Plasma, oxidized, DiI conjugate (DiI-OxLDL), 200 μL - FAQs
Low Density Lipoprotein from Human Plasma, oxidized, DiI conjugate (DiI-OxLDL), 200 μL - FAQs
View additional product information for Oxidized Low-density Lipoprotein (OxLDL) and DiI-OxLDL - FAQs (L34358, L34357)
2 product FAQs found
If the Low Density Lipoprotein (LDL) from Human Plasma was accidentally frozen, can I still use it?
We recommend storing Low Density Lipoprotein (LDL) from Human Plasma at 2-8 degrees C. We do not recommend using frozen product.
Can I use Low Density Lipoprotein (LDL) from Human Plasma on other species?
We have not tested this.