LIVE/DEAD™ Fixable Olive (557) Viability Kit, for 488 nm excitation, 400 Assays - FAQs
LIVE/DEAD™ Fixable Olive (557) Viability Kit, for 488 nm excitation, 400 Assays - FAQs
View additional product information for LIVE/DEAD™ Fixable Olive (557) Viability Kit, for 488 nm excitation - FAQs (L34979, L34978, L34977)
1 product FAQs found
What is the concentration of the LIVE/DEAD Fixable dyes?
The concentration of LIVE/DEAD Fixable dyes is proprietary.