LIVE/DEAD™ Fixable Scarlet (723) Viability Kit, for 633 nm excitation, 400 Assays - FAQs
LIVE/DEAD™ Fixable Scarlet (723) Viability Kit, for 633 nm excitation, 400 Assays - FAQs
View additional product information for LIVE/DEAD™ Fixable Scarlet (723) Viability Kit, for 633 nm excitation - FAQs (L34988, L34987, L34986)
1 product FAQs found
What is the concentration of the LIVE/DEAD Fixable dyes?
The concentration of LIVE/DEAD Fixable dyes is proprietary.