If the Low Density Lipoprotein (LDL) from Human Plasma was accidentally frozen, can I still use it?
We recommend storing Low Density Lipoprotein (LDL) from Human Plasma at 2-8 degrees C. We do not recommend using frozen product.