LysoLight™ Antibody Labeling Kits
Invitrogen™

LysoLight™ Antibody Labeling Kits

The LysoLight Antibody Labeling Kits offer an efficient method for conjugating antibodies with LysoLight dyes. This process allows for specific and sensitive detection of internalized antibodies within lysosomes using fluorescence microscopy, flow cytometry, or high content imaging platforms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct TypeColor
L360023 x 1 mgAntibody Labeling KitDeep Red
L360013 x 100 μgAntibody Labeling KitDeep Red
L360033 x 100 μgAmine Reactive DyeDeep Red
L36004500 μgAmine Reactive DyeDeep Red
L360053 x 100 μgAntibody Labeling KitGreen
L360063 x 1 mgAntibody Labeling KitGreen
L360073 x 100 μgAmine Reactive DyeGreen
L36008500 μgAmine Reactive DyeGreen
Catalog number L36002
Price (USD)
722.00
Each
Quantity:
3 x 1 mg
Product Type:
Antibody Labeling Kit
Color:
Deep Red
Price (USD)
722.00
Each
LysoLight Antibody Labeling Kits provide a powerful tool for visualizing the lysosomal localization and catabolic degradation of antibodies and proteins. These kits utilize a unique fluorophore that is only active upon enzymatic cleavage by the lysosomal specific enzyme, Cathepsin B. Outside of the lysosome, LysoLight dyes have little to no observable background fluorescence, and only upon localization to the protease rich environment of the lysosome, does LysoLight become activated and brightly fluoresce bright green or deep red in color.
The labeling protocol provided in the LysoLight Antibody Labeling Kits involves a simple and rapid procedure, ensuring minimal hands-on time and reliable results. The kits include all necessary reagents, including the LysoLight dye, buffers, and purification columns, to facilitate the antibody labeling process. The LysoLight reactive dye contained in these kits readily reacts with lysine residues in antibodies or proteins to yield a covalently attached fluorogenic sensor. The spin columns included in the kits are used for purifying the labeled antibody from excess dye with yields of 70–95%. The kits contain sufficient reagents for three labeling reactions of 100 μg or 1 mg of antibody. The user guide describes a general protocol for using the amine-reactive form of the LysoLight dye for labeling either 100 μg or 1 mg of IgG.
For Research Use Only.
Specifications
DescriptionLysoLight™ Deep Red Antibody Labeling Kit
Detection MethodFluorescence
Excitation/Emission650/668 nm
Quantity3 x 1 mg
SolubilityDMSO (Dimethylsulfoxide)
ColorDeep Red
For Use With (Application)Antibody and Protein Labeling
For Use With (Equipment)Flow Cytometer, Fluorescence Microscope, High Content Analysis Instrument
Product LineLysoLight
Product TypeAntibody Labeling Kit
Unit SizeEach