What is the recommended incubation time for LysoTracker Green DND-26 (Cat. No. L7526) and LysoSensor Yellow/Blue DND-160 (Cat. No. L7545)?
LysoTracker Green DND-26 (Cat. No. L7526) and LysoSensor Yellow/Blue DND-160 (Cat. No. L7545) show faster uptake in cells and consequently a faster "alkalizing effect" on the lysosomes, so we recommend incubating the cells with these probes for 1-5 min at 37 degrees C. As the other LysoTracker and LysoSensor probes have a slower rate of uptake, we recommend a longer incubation time of 30 min to 2 hr.