Gel Loading Tips, standard, round
Invitrogen™

Gel Loading Tips, standard, round

Gel Loading Tips (standard, round) can be used for easy and accurate loading of samples onto 1.0 mm and 1.5Read more
Have Questions?
Catalog NumberQuantity
LC1001200/ pkg.
Catalog number LC1001
Price (USD)
95.65
Online exclusive
100.00
Save 4.35
Each
Quantity:
200/ pkg.
Price (USD)
95.65
Online exclusive
100.00
Save 4.35
Each
Gel Loading Tips (standard, round) can be used for easy and accurate loading of samples onto 1.0 mm and 1.5 mm gels. The loading volume for these tips is 0.5–200 μL. These tips are RNase/DNase-free and non-autoclavable.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionStandard round Tips
FilteredNo
Quantity200/ pkg.
Shipping ConditionRoom Temperature
SterilityNon-sterile
Volume (Metric)200 μL
For Use With (Application)1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
For Use With (Equipment)Universal Fit
TypeGel Loading Tip
Unit SizeEach
Contents & Storage
Contains 1 pack (200/pack) of Standard, Round gel loading tips.

Frequently asked questions (FAQs)

Do you offer gel loading tips to load protein gels?

We offer Invitrogen Gel Loading Tips for easy and accurate loading of samples. They are RNase/DNase-free and non-autoclavable. Three varieties are offered: standard round tips, flat gel tips, and Eppendorf round gel tips.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.