Invitrogen offers a variety of tips for easy and accurate loading of samples. They are RNase/DNase-free and non-autoclavable.Read more
Have Questions?
Catalog Number
Quantity
LC1002
200/ pkg.
Catalog number LC1002
Price (USD)
167.65
Online exclusive
175.00
Save 7.35
Each
Quantity:
200/ pkg.
Price (USD)
167.65
Online exclusive
175.00
Save 7.35
Each
Invitrogen offers a variety of tips for easy and accurate loading of samples. They are RNase/DNase-free and non-autoclavable.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionFlat Tips
FilteredNo
Quantity200/ pkg.
Shipping ConditionRoom Temperature
SterilityNon-sterile
Volume (Metric)200 μL
For Use With (Application)1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
For Use With (Equipment)Universal Fit
Product LineEppendorf
TypeFlat Gel Loading Tip
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
Do you offer gel loading tips to load protein gels?
We offer Invitrogen Gel Loading Tips for easy and accurate loading of samples. They are RNase/DNase-free and non-autoclavable. Three varieties are offered: standard round tips, flat gel tips, and Eppendorf round gel tips.