Invitrolon™ PVDF/Filter Paper Sandwiches, 0.45 μm, 8.3 x 7.3 cm (for mini gels)
Invitrogen™

Invitrolon™ PVDF/Filter Paper Sandwiches, 0.45 μm, 8.3 x 7.3 cm (for mini gels)

Invitrolon PVDF/Filter Paper Sandwiches, 0.45 μm, have excellent binding properties for western blotting, dot-blot assays, and protein sequencing. The 0.45-μmRead more
Have Questions?
Catalog NumberQuantity
LC200520 membrane/filter paper sandwiches
Catalog number LC2005
Price (USD)
333.00
Each
Quantity:
20 membrane/filter paper sandwiches
Price (USD)
333.00
Each
Invitrolon PVDF/Filter Paper Sandwiches, 0.45 μm, have excellent binding properties for western blotting, dot-blot assays, and protein sequencing. The 0.45-μm pore size makes these membranes ideal for protein transfer for proteins >10kDa.

See all membranes and filter papers ›

Features
• Membrane: polyvinylidene difluoride (PVDF)
• Filter paper thickness: 0.8 mm
• Binding capacity: goat IgG: 294 μg/cm2; BSA: 131 μg/cm2; insulin: 85 μg/cm2
• Re-probe characteristics: yes
• Pre-activation: required with 100% alcohol (methanol, ethanol, or isopropanol)
• Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
• Durability: compatible with most organic solvents, acids, and mild bases
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity20 membrane/filter paper sandwiches
Shipping ConditionRoom Temperature
Dimensions (LxW)7.3 cm x 8.3 cm
FormatSandwich
Length (Metric)8.3 cm
MaterialPVDF
Pore Size0.45 μm
Product LineInvitrolon Novex
Thickness0.8 mm
Width (Metric)7.3 cm
Unit SizeEach
Contents & Storage
Invitrolon™ PVDF are conveniently supplied as 20 membrane/filter paper sandwiches per pack. Store at room temperature.

Frequently asked questions (FAQs)

How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why must Invitrolon PVDF membranes be dried before staining?

Drying the PVDF membrane reduces the background staining that can occur with wet membranes. A dry PVDF membrane is very hydrophobic and doesn't wet well, but the areas where proteins are bound are more easily saturated and stainable.

This will not work with nitrocellulose membranes, and will only work with PVDF membranes stained for a brief period; staining beyond the recommended time will only increase the background and reduce the detectability.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How do I destain proteins on a PVDF membrane that were stained with SimplyBlue SafeStain?

After staining with SimplyBlue SafeStain, use deionized water for the less strongly retained protein bands on the PVDF membrane.

Increasing methanol or ethanol concentrations up to 70% should destain any remaining bands. You can leave the membrane in the destain indefinitely.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I use ethanol instead of methanol in the NuPAGE transfer buffer (and in transfer buffers in general)?

We find that including ethanol in the transfer buffer is just as effective as including methanol. You may use either of these alcohols at an equivalent concentration.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How can I reduce high background on PVDF Western blots?

PVDF membranes require more stringent blocking steps. This can be achieved by increasing the concentration of the blocking reagent 2 to 5 fold, increasing the blocking time, and performing the procedure at 37 degrees C. Blocking agents bind to unoccupied sites to prevent background staining and also to membrane-bound proteins, thus reducing non-specific interactions with the primary antibody. We offer WesternBreeze Immunodetection kits with blocking reagents and Pierce Fast Western Blot kits that have been pre-optimized to give low-background blots. Other examples of blocking agents are nonfat dry milk, BSA, and casein, Starting Block, and SuperBlock.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Citations & References (3)

Citations & References
Abstract
Molecular cloning and characterization of a distinct human phosphodiesterase gene family: PDE11A.
Authors:Fawcett L, Baxendale R, Stacey P, McGrouther C, Harrow I, Soderling S, Hetman J, Beavo JA, Phillips SC
Journal:Proc Natl Acad Sci U S A
PubMed ID:10725373
We report here the cloning, expression, and characterization of human PDE11A1, a member of a distinct cyclic nucleotide phosphodiesterase (PDE) family. PDE11A exhibits </=50% amino acid identity with the catalytic domains of all other PDEs, being most similar to PDE5, and has distinct biochemical properties. The human PDE11A1 cDNA isolated ... More
Protein kinase C epsilon-dependent regulation of cystic fibrosis transmembrane regulator involves binding to a receptor for activated C kinase (RACK1) and RACK1 binding to Na+/H+ exchange regulatory factor.
Authors: Liedtke Carole M; Yun C H Chris; Kyle Nicole; Wang Dandan;
Journal:J Biol Chem
PubMed ID:11956211
Protein kinase C (PKC) regulation of cystic fibrosis transmembrane regulator (CFTR) chloride function has been demonstrated in several cell lines, including Calu-3 cells that express native, wild-type CFTR. We demonstrated previously that PKC epsilon was required for cAMP-dependent CFTR function. The goal of this study was to determine whether PKC ... More
EMR4, a Novel Epidermal Growth Factor (EGF)-TM7 Molecule Up-regulated in Activated Mouse Macrophages, Binds to a Putative Cellular Ligand on B Lymphoma Cell Line A20.
Authors: Stacey Martin; Chang Gin-Wen; Sanos Stephanie L; Chittenden Laura R; Stubbs Lisa; Gordon Siamon; Lin Hsi-Hsien;
Journal:J Biol Chem
PubMed ID:12023293
A novel member of the EGF-TM7 family, mEMR4, was identified and characterized. The full-length mouse EMR4 cDNA encodes a predicted 689-amino acid protein containing two epidermal growth factor (EGF)-like modules, a mucin-like spacer domain, and a seven-transmembrane domain with a cytoplasmic tail. Genetic mapping established that mEMR4 is localized in ... More