Blotting Roller - FAQs
Blotting Roller - FAQs
View additional product information for Blotting Roller - FAQs (LC2100)
1 product FAQs found
What method do you recommend for blotting E-PAGE gels?
The preferred method of transfer for E-PAGE gels is dry blotting using the iBlot/iBlot 2 Dry Blotting System (see Page 27 of the E-PAGE Technical Guide http://tools.thermofisher.com/content/sfs/manuals/epagetechguide_man.pdf). Please note that this works only for E-PAGE 48 gels. Semi-dry blotting using the Invitrogen Semi-Dry Blotter, Cat. No. SD1000 (Page 33), or semi-wet blotting (Page 37) may also be performed.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.