Novex™ Zymogram Developing Buffer (10X) - FAQs
Novex™ Zymogram Developing Buffer (10X) - FAQs
View additional product information for Novex™ Zymogram Developing Buffer (10X) - FAQs (LC2671)
2 product FAQs found
Does the Colloidal Blue Staining kit work with Invitrogen Zymogram gels?
Yes, the Colloidal Blue Staining kit works very well with Invitrogen Zymogram gels.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the Brij 35 detergent concentration in the 1X Zymogram Developing buffer?
The Brij 35 detergent concentration for Zymogram Developing buffer should be 0.02% (w/v).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.