Novex™ Tris-Glycine SDS Running Buffer (10X), 4 x 1 L - FAQs
Novex™ Tris-Glycine SDS Running Buffer (10X), 4 x 1 L - FAQs
View additional product information for Novex™ Tris-Glycine SDS Running Buffer (10X) - FAQs (LC2675, LC26754, LC26755)
1 product FAQs found