Novex™ Hi-Density TBE Sample Buffer (5X)
Invitrogen™

Novex™ Hi-Density TBE Sample Buffer (5X)

Novex™ TBE Gels provide the highest resolution possible for analyzing restriction digests and PCR products. DNA fragments ranging from 10Read more
Have Questions?
Catalog NumberQuantity
LC667810 mL
Catalog number LC6678
Price (USD)
158.00
Each
Quantity:
10 mL
Request bulk or custom format
Price (USD)
158.00
Each
Novex™ TBE Gels provide the highest resolution possible for analyzing restriction digests and PCR products. DNA fragments ranging from 10 to 3,000 base pairs are clearly resolved into sharp, tight bands. Novex™ DNA Retardation gels are prepared with 0.5X TBE as the gel buffer. This is sufficient for good electrophoretic separation, yet low enough to promote DNA-protein interactions. Novex™ TBE and DNA Retardation Gels are designed to run on the XCell SureLock™ Mini-Cell.

Formulation:

Novex™ TBE and DNA Retardation Gels are manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS.

Recommended Buffers:

For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex™ Hi-Density TBE Sample Buffer contains the density agent Ficoll™, which yields sharper, straighter bands than conventional density agents, and the tracking dyes Bromophenol Blue and Xylene Cyanol.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffer
Concentration5 X
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting
Gel CompatibilityPolyacrylamide Gels
Label or DyeBromophenol Blue, Xylene Cyanol
Product LineNovex
Product TypeTBE Buffer
Quantity10 mL
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C

Frequently asked questions (FAQs)

My Novex TBE buffer has precipitated out of solution. What can I do?

If a slight turbidity develops, the fine precipitate can be dissolved by autoclaving for 5 minutes at 110°C. Do not autoclave in the container supplied. This treatment has no deleterious effect on the buffering properties of TBE.

Do you have the recipe for the TBE running buffer and the Hi-Density TBE Sample Buffer?

Please see the recipes below:

TBE Running Buffer (5X):
Tris base, 54.0 g
Boric acid, 27.5 g
EDTA (free acid), 2.9 g
Deionized water to 1.0 L

Hi-Density TBE Sample Buffer (5X):
5X TBE Running buffer (Cat. No. LC6675), 2 mL
Ficoll Type 400, 1.5 g
Bromophenol blue, 1 mL of 1% solution
Xylene cyanol 1 mL of 1% solution
Deionized water to 10.0 mL