CapSure™ Macro LCM Caps, 240 Caps - FAQs
CapSure™ Macro LCM Caps, 240 Caps - FAQs
View additional product information for CapSure™ Macro LCM Caps - FAQs (LCM0212, LCM0211)
1 product FAQs found
Are CapSure LCM Caps compatible with instruments other than ArcturusXT LCM instruments?
CapSure LCM Caps are specifically designed for use with Arcturus LCM instruments. Therefore, we do not recommend using them with other instruments.