CapSure™ HS LCM Caps (Alignment Tray & Incubation Block required, not included), 160 Caps - FAQs
CapSure™ HS LCM Caps (Alignment Tray & Incubation Block required, not included), 160 Caps - FAQs
View additional product information for CapSure™ HS LCM Caps (Alignment Tray & Incubation Block required, not included) - FAQs (LCM0214, LCM0215)
1 product FAQs found
Are CapSure LCM Caps compatible with instruments other than ArcturusXT LCM instruments?
CapSure LCM Caps are specifically designed for use with Arcturus LCM instruments. Therefore, we do not recommend using them with other instruments.