Is the LCM0522 PEN Membrane Glass Slide compatible with Leica LMD7 systems?
The LCM0522 PEN Membrane Glass Slide is designed for use with Arcturus LCM instruments, including the Arcturus Cellect and Accuva Cellect systems. Compatibility with Leica LMD7 systems has not been established. For specific guidance, contact technical support.
How should I prepare slides for LCM?
The Arcturus HistoGene LCM Frozen Section Staining Kit (Cat. No. KIT0401) comes complete with all the reagents and supplies needed for preparing frozen tissue sections for laser capture microdissection (LCM). Designed as a fast-penetrating stain, this kit provides excellent contrast while preserving nucleic acid integrity and retaining low-abundance mRNAs.
The Arcturus Paradise Plus FFPE LCM Staining Kit (Cat. No. KIT0312J) offers a simple solution to stain formalin-fixed, paraffin-embedded (FFPE) tissues before proceeding with laser capture microdissection (LCM) and downstream molecular analysis. FFPE samples introduce unique challenges for gene expression profiling, including chemical modification and fragmentation of RNA molecules. Archiving of FFPE samples adds to these challenges through increased RNA degradation over time.
The Arcturus HistoGene LCM Immunofluorescence Staining Kit (Cat. No. KIT0420) is designed to retrieve high-quality nucleic acids from immunofluorescently stained, frozen tissue using a quick 15-minute process. The kit provides reagents and slides for convenient and reliable immunofluorescent staining as well as protocols that are streamlined and optimized to maintain nucleic acid quality. The staining kit is used with a biotinylated monoclonal primary antibody against an antigen of choice provided by the user, and Cyanine 3–conjugated streptavidin is included in the kit.