PLCG1 Monoclonal Antibody (1F1) - FAQs
PLCG1 Monoclonal Antibody (1F1) - FAQs
View additional product information for PLCG1 Monoclonal Antibody (1F1) - FAQs (LFMA0049)
1 product FAQs found
Do you offer antibodies for PLC-g1?
Yes, we have several monoclonals against PLC-g1 (Cat. Nos. 700044, LF-MA0050, LF-MA0049) and polyclonal antibodies against phosho-forms of PLC-g1 (Cat. Nos.44-696G, PA538090, and PA538318, among others).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.