Biomarker 3-Plex Human PlexMark™ Panel, Polystyrene - FAQs
Biomarker 3-Plex Human PlexMark™ Panel, Polystyrene - FAQs
View additional product information for Biomarker 3-Plex Human PlexMark™ Panel, Polystyrene - FAQs (LHC6007)
3 product FAQs found
Is it possible to reduce sample volumes for the analysis of extracellular proteins in multiplex immunoassay for the Luminex platform?
Reduction of the sample volume by half, 25 uL, is a viable alternative when sample volume is limiting.