PIM-1 [pS8] Singleplex Bead Kit - FAQs
PIM-1 [pS8] Singleplex Bead Kit - FAQs
View additional product information for PIM-1 [pS8] Singleplex Bead Kit - FAQs (LHO6571)
3 product FAQs found
Is it possible to reduce sample volumes for the analysis of extracellular proteins in multiplex immunoassay for the Luminex platform?
Reduction of the sample volume by half, 25 uL, is a viable alternative when sample volume is limiting.