Search
View additional product information for Caspase-3 [175/ 176] Singleplex Bead Kit - FAQs (LHO6581)
3 product FAQs found
Reduction of the sample volume by half, 25 uL, is a viable alternative when sample volume is limiting.