TRACE™ 1610 Li-ion Battery Gas Analyzer
TRACE™ 1610 Li-ion Battery Gas Analyzer
TRACE™ 1610 Li-ion Battery Gas Analyzer
Thermo Scientific™
TRACE™ 1610 Li-ion Battery Gas Analyzer
Analyze Li-ion battery off-gases with automated Thermo Scientific™ TRACE™ 1610 GC gas analyzer, for electrolytes decomposition study and battery failure detection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
LIBATAN1610-230230 V
LIBATAN1610-120120 V
Catalog number LIBATAN1610-230
Price (USD)
-
Voltage:
230 V
Reliably detect Li-ion battery outgassing with the Thermo Scientific™ TRACE™ 1610 Li-ion Battery Gas Analyzer, and get important information about possible deterioration and performance of different battery formulations.
Initial charge/discharge processes in batteries produce a variety of gas components that have an impact on battery performance after long-term use. Additionally, some flammable gases create a security risk. This multi-column, multi-valve gas chromatography solution offers a reliable tool for the analysis of gases causing lithium battery swelling.
Gas composition produced by batteries is highly complex. There are three categories:
  • Permanent gases such as hydrogen, methane, carbon monoxide, carbon dioxide
  • Short-chain hydrocarbons (C2–C5)
  • Other volatile compounds
The GC analyzer is delivered fully tested and includes a complete documentation package with method, plumbing diagram, setup and troubleshooting guide, and spare parts and consumables list.
Data system and reporting package are not included but available as separate items.
Built to ensure robustness and analytical performance
  • Spacious independent auxiliary oven with easy access to valves, rotors, and actuators
  • GC features iConnect injector and detector modules facilitating troubleshooting and maintenance
  • Optimized system footprint to reduce benchspace requirement
Specifications
Dimensions (L x W x H)67 x 75 x 45 cm (26.4 x 29.5 x 17.7 in.)
FunctionAnalysis of Li-ion battery swelling gas composition
IncludesGas chromatograph, split/splitless inlet, TCD, FID, auxiliary valve oven, two 6-port valves, auxiliary gas module, columns, and method
Inlet TypeSSL
ModelGC Analyzer for Lithium-ion Battery Swelling Gas
Voltage230 V
Column Capacity3 (2 Packed, 1 Capillary)
Detector TypeTCD, FID
Product TypeLi-Ion Battery Gas Analyzer
Unit SizeEach