Oxoid™ Lab-Lemco Beef Extract Powder
Actual product may vary

Oxoid™ Lab-Lemco Beef Extract Powder

Thermo Scientific™ Oxoid Lab-Lemco Powder is made with premium beef sources to deliver a nutritious and safe product.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0029T5 kg
LP0029B500 g
LP0029K25 kg
LP0029G10 kg
Catalog number LP0029T
Price (USD)
-
Quantity:
5 kg
Thermo Scientific™ Oxoid Lab-Lemco Powder is a Beef Extract made from specially selected raw materials for quality and bio-logical safety.

Use with other refined ingredients, e.g. Neutralized bacteriological peptone (LP0034) to prepare culture media which do not require filtration.

May be used in fermentation processes.

Industrial Reference: NMKL

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Fermentation processes
Protein SourceBeef
Quantity5 kg
Shelf Life720 Days from Manufacture
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage