Oxoid™ Casein Hydrolysate
Thermo Scientific™

Oxoid™ Casein Hydrolysate

Oxoid Casein Hydrolysate (Acid) is a purified hydrolysate high in nitrogenous material ideal for culture media or as an amino acid mixture.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0041B500 g
LP0041T5 kg
Catalog number LP0041B
Price (USD)
-
Quantity:
500 g
Thermo Scientific™ Oxoid Casein Hydrolysate (Acid) is a purified hydrolysate which provides the necessary nitrogenous material for culture media or can be used in fermentation when amino acid mixtures are specified.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
DescriptionDehydrated; Casein hydrolysate (Acid)
For Use With (Application)Culture media or fermentation processes
Protein SourceCasein
Quantity500 g
Shelf Life720 Days from Manufacture
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage