Oxoid™ Tryptone
Oxoid™ Tryptone
Thermo Scientific™

Oxoid™ Tryptone

The Thermo Scientific™ Oxoid Tryptone is a high performance pancreatic digest of casein.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0042T5 kg
LP0042B500 g
LP0042R2.5 kg
LP0042K25 kg
Catalog number LP0042T
Price (USD)
-
Request A Quote
Quantity:
5 kg
The Thermo Scientific™ Oxoid Tryptone ia a pancreatic digest of casein for use where high performance and uniformity of composition are of paramount importance.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Culture media or fermentation processes
Protein SourceCasein
Quantity5 kg
Shelf Life5 years
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage