Oxoid™ Bile Salts
Oxoid™ Bile Salts
Thermo Scientific™
Oxoid™ Bile Salts
Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0055T5 kg
LP0055J250 g
Catalog number LP0055T
Price (MXN)
-
Request A Quote
Quantity:
5 kg
Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.
Normal working concentration 0.5% w/v. Each vial supplements 500 mL of medium.
Specifications
Quantity5 kg
FormPowder
Product TypeBile Salts
Unit SizeEach