This product will be discontinued once remaining inventory is depleted. We offer the IL-8 Monkey Magnetic Kit for Luminex™ Platform as a replacement.

IL-8 Monkey Singleplex Kit

Catalog number: LPC0081

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Singleplex bead kit comprises the analyte specific components for the measurement of monkey IL-8 (Interleukin-8) in serum, plasma or tissue culture supernatant. The assay may be run alone or in combination with other Monkey Singleplex Bead Kits. These reagents are intended for use with the Luminex™ 100™/200™ and FLEXMAP 3D™ systems. Buffer reagents needed to complete the reaction are sold separately (Cat. No. LHB0001). The Invitrogen EveryPrep™ Universal Vacuum Manifold (Cat. No. K2111-01) is available.

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: See product documentation
Assay Sensitivity: 15 pg/mL
Bead Region: 81
Former Identifiers: Novex
Gene Name: C-X-C motif chemokine ligand 8
Gene Symbol: CXCL8
Immunoassay Kit Format: Singleplex Kit
Protein Aliases: IL-8, C-X-C motif chemokine 8, Chemokine (C-X-C motif) ligand 8
Protein Name: Interleukin-8
Target (Kit Named): IL-8
Target Species (Named): Monkey
Target Species (Validated): Monkey
UniProt ID: P67813
Detection Method: Fluorescent
For Use With (Equipment): Luminex™ Instruments
Gene Aliases: IL8
Gene ID: 613028
Label or Dye: R-PE
Product Size: 100 tests
Sample Type (Specific): Cell Culture Supernatants, Plasma, Serum
System: Luminex™
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8°C.

Documents

Manuals & protocols