Need free, one-on-one consultation on running your assay? For help, email luminexFAS@lifetech.com.

CXCL10 Monkey Singleplex Kit

Catalog number: LPC1171M

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This singleplex bead kit is specifically designed for the measurement of IP-10 (Interferon-gamma-inducible, 10 kD protein) in Rhesus monkey and Cynomolgus monkey serum, plasma, and tissue culture supernatant. Assays using the IP-10 Monkey Singleplex Magnetic Bead Kit may be run alone or in combination with the Monkey Cytokine Magnetic 28-Plex Panel. This singleplex bead kit employs magnetic beads, facilitating automation, decreasing hands-on time, and increasing throughput and precision. The panel is suitable for use with the Luminex™ 200™, FLEXMAP 3D™, and MAGPIX™ systems. Buffer reagents needed to complete the reaction are sold separately (Catalogue number LHB0001M).
Superior performance—accurate, reproducible, and sensitive quantitation of selected proteins

• High quality
—fully qualified antibodies permit excellent specificity and sensitivity

• Fast and easy protocols
—perform your assays and analyze your data typically in less than one day

Luminex™ xMAP™ Technology—Effective and Proven Analytical Tool
The IP-10 Monkey Singleplex Magnetic Bead Kit is based on xMAP™ technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex™ assays. Magnetic microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead is given a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format. Upon completion of the sandwich immunoassay, magnetic beads must be measured using a Luminex™ detection system (MAGPIX™, 100™/200™, or FLEXMAP 3D™). The instrument uses xPONENT™ software to distinguish bead color (analyte) and R-PE fluorescence intensity (assay signal strength) to quantify target(s).

Magnetic Beads Let You Do More with Your Time
Magnetic bead-based assays use Luminex™ MagPlex magnetic microsphere. All other assay components are the same as the equivalent Luminex™ polystyrene bead-based assays providing identical quality and consistency performance. MagPlex technology takes advantage of magnetic properties to simplify assay wash steps and maximize uniformity of results. Magnetic bead-based assays also enable automation, decrease hands-on time, and increase throughput and precision. Easy to use protocols allow you to get your results in as little as 3.5 hours. In addition, these assays are compatible with both vacuum and magnetic wash stations and work well with the MAGPIX™ system as well as other Luminex™ xMAP™ platforms.

More Validation for Confidence in Your Results
The IP-10 Monkey Singleplex Magnetic Bead Kit has been validated using the same rigorous criteria that distinguishes Luminex™ assays from other commercially available assays. With more than 20 years of experience, our fully qualified antibodies help ensure the specificity and sensitivity of our assays. Every singleplex kit and premixed multiplex kit comes with a product insert that outlines the specifications for each marker in the assay (see Documents section below).

Learn more about all Invitrogen™ assays for the Luminex™ platform.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: See product documentation
Assay Sensitivity: See product documentation
Bead Region: See product documentation
Former Identifiers: Novex
Gene Name: C-X-C motif chemokine ligand 10
Gene Symbol: CXCL10
Immunoassay Kit Format: Singleplex Kit
Protein Aliases: 10 kDa interferon gamma-induced protein, Gamma-IP10, IP-10, Small-inducible cytokine B10
Protein Name: C-X-C motif chemokine 10
Target (Kit Named): CXCL10
Target Species (Named): Monkey
Target Species (Validated): Monkey
UniProt ID: Q8MIZ1
Detection Method: Fluorescent
For Use With (Equipment): Luminex™ Instruments
Gene ID: 574243
Label or Dye: R-PE
Product Size: 100 tests
Sample Type (Specific): Serum
System: Luminex™
Shipping Condition: Wet Ice

Contents & storage

Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet. Store at 2-8° C.

Documents

Manuals & protocols