Cartridges for Multidrop™ Pico Digital Dispensers
Thermo Scientific™
Cartridges for Multidrop™ Pico Digital Dispensers
Cartridges for Multidrop™ Pico Digital Dispensers allow dispensing of one to eight fluids per cartridge, from 8 pL to 10 μL of DMSO, aqueous fluids, or master mixes, with or without small molecules or biomolecules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ChannelsModel
LTR000411 pL to 10 μL88-channel 20 μL Standard Cartridges
LTR000111 pL to 10 μL11-channel 20 μL Cartridges
LTR00021 nL to 10 μL11-channel 200 μL Cartridges
LTR00031 nL to 10 μL44-channel 200 μL Standard Cartridges
LTR00051 nL to 10 μL4Surfactant Free Cartridges, 4-channel 200 μL
LTR00068 pL to 10 μL8Surfactant Free Cartridges, 8-channel 20 μL
Catalog number LTR0004
Price (USD)
1,401.30
Online exclusive
1,557.00
Save 155.70
Case of 20
Volume (Metric):
11 pL to 10 μL
Channels:
8
Model:
8-channel 20 μL Standard Cartridges
Price (USD)
1,401.30
Online exclusive
1,557.00
Save 155.70
Case of 20
The cartridges are designed for use with Multidrop™ Pico Digital Dispensers, dispensing volumes from 8 pL to 10 μL. Use the 1-channel cartridges with the Multidrop™ Pico 1, and the 4- or 8-channel standard or surfactant free cartridges with the Multidrop™ Pico 8 Surfactant Free (Pico 8 sf).
Features:
  • Single-Channel Cartridges: Use the 1-channel cartridges with the Pico 1 to dispense DMSO, aqueous fluids with added surfactant, or master mixes. Dispensing volume: 11 pL – 10 μL..
  • Standard Cartridges: Use the 4- or 8-channel standard cartridges with the Pico 8 sf to dispense DMSO, aqueous fluids with added surfactant, or master mixes. Dispensing volume: 11 pL – 10 μL.
  • Surfactant Free Cartridges: Use the 4- or 8-channel surfactant free cartridges with the Pico 8 sf to dispense only aqueous fluids without surfactant. Dispensing volume: 8 pL – 10 μL.
  • Contamination-Free Dispensing: Non-contact dispensing and single-use cartridges prevent carry-over and contamination.
Specifications
For Use With (Equipment)Multidrop Pico 8 Digital Dispenser, Multidrop Pico 8 Surfactant Free Digital Dispenser
Model8-channel 20 μL Standard Cartridges
No. per PackCase of 20
Channels8
Product TypeDigital Dispenser Cartridge
Volume (Metric)11 pL to 10 μL
Unit SizeCase of 20