I am looking for the sequence information and epitope for MUC5AC Monoclonal Antibody (2-25LE) (Cat. No. MA1-19346)?
This antibody was prepared against the full-length mucin protein, isolated from an ovarian cyst fluid. Typically, for monoclonal antibodies, the immunogen is long and the exact location of binding is not known. We do not systematically test for the exact epitope for our primary antibodies, as this would require an extensive experimental project for each one.
For more information, the following link is for a book regarding epitope mapping: https://doi.org/10.1007/978-1-4939-7841-0.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
For more information, the following link is for a book regarding epitope mapping: https://doi.org/10.1007/978-1-4939-7841-0.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.