Cas9 Monoclonal Antibody (10C11-A12) - FAQs
Cas9 Monoclonal Antibody (10C11-A12) - FAQs
View additional product information for Cas9 Monoclonal Antibody (10C11-A12) - FAQs (MA1202)
1 product FAQs found
Is there a specific antibody that you recommend to test the delivery of TrueCut Cas9 Protein v2?
Although we do not have a specific antibody recommendation to test the delivery of TrueCut Cas9 Protein v2, we have tested the following antibodies against an older version of Cas9 (Platinum Cas9) using U2OS-Cas9 expression cell lines (generated using Cas9 lentivirus):
- Cas9 Recombinant Polyclonal Antibody (Cat. No. 711061)
- Cas9 Monoclonal Antibody (Cat.No. MA1-201)
- Cas9 Monoclonal Antibody (Cat. No. MA1-202)
Our TrueCut Cas9 Protein v2 and the Platinum Cas9 have similar domains, and the TrueCut Cas9 Protein v2 has better efficiencies. Therefore, these antibodies should work with TrueCut Cas9 Protein v2 as well.