Mouse IgG1 Isotype Control (MOPC-21), Alexa Fluor™ 488 - FAQs
Mouse IgG1 Isotype Control (MOPC-21), Alexa Fluor™ 488 - FAQs
View additional product information for Mouse IgG1 Isotype Control (MOPC-21), Alexa Fluor™ 488 - FAQs (MA518167)
1 product FAQs found
What is the shelf life of Mouse IgG1 Isotype Control (MOPC-21), Alexa Fluor 488 (Cat. No. MA5-18167)?
Mouse IgG1 Isotype Control (MOPC-21), Alexa Fluor 488 is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.