What is the antibody concentration for MLKL Recombinant Rabbit Monoclonal Antibody (2B9) (Cat. No. MA5-24846)?
MLKL Recombinant Rabbit Monoclonal Antibody (2B9) (Cat. No. MA5-24846) has a concentration of ~1 mg/mL. Slight variation from this may be expected due to lot-to-lot variation and this information will be received with the product.