AMACR Monoclonal Antibody (13H4) - FAQs
AMACR Monoclonal Antibody (13H4) - FAQs
View additional product information for AMACR Monoclonal Antibody (13H4) - FAQs (MA514576)
1 product FAQs found
What is the concentration of AMACR Monoclonal Antibody (13H4) (Cat. No. MA514576)?
This antibody is tested according to its approved application of IHC-FFPE to ensure proper performance. However, its concentration is not quantified.