What was the pH of the buffer used for validating the IGHA1 Monoclonal Antibody (7D5F12) (Cat. No. MA5-31774)?
The pH of the buffer used for validating the IGHA1 Monoclonal Antibody (Cat. No. MA5-31774) was pH 6.
Search
View additional product information for IGHA1 Monoclonal Antibody (7D5F12) - FAQs (MA531774)
1 product FAQs found