BenchPro™ 2100 Air Compressor, 220 Volt - FAQs
BenchPro™ 2100 Air Compressor, 220 Volt - FAQs
View additional product information for BenchPro™ 2100 Air Compressor, 220 Volt - FAQs (MC5001220)
1 product FAQs found
How much alcohol is in the BenchPro 2100 Plasmid Purification Card and Reagent Tray Kit?
There are 5.58mL of isopropanol and 5.33mL of 70% ethanol per tray in the BenchPro 2100 Plasmid Purification Card and Reagent Tray Kit.