CD25 Monoclonal Antibody (CD25-3G10), PE-Alexa Fluor™ 700 - FAQs
CD25 Monoclonal Antibody (CD25-3G10), PE-Alexa Fluor™ 700 - FAQs
View additional product information for CD25 Monoclonal Antibody (CD25-3G10), PE-Alexa Fluor™ 700 - FAQs (MHCD2524)
1 product FAQs found
What is the concentration of CD25 Monoclonal Antibody (CD25-3G10), PE-Alexa Fluor 700 (Cat. No. MHCD2524)?
The concentration of this antibody is lot-dependent and can be found in the Certificate of Analysis for that lot number.