TriPlus™ 500 GC Headspace Autosampler
Thermo Scientific™
TriPlus™ 500 GC Headspace Autosampler
Boost productivity and data quality during volatiles analysis with the highly reliable, robust, and compact TriPlus 500 Headspace Autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeDescription
MI-25118000Vial Loader and Tray UpgradeUpgrade to 120 vial capacity
25118200Vial Tray UpgradeUpgrade to 240 vial capacity
MI-25118100Headspace Autosamplerwith Direct Column Connection
MI-25118300Headspace Autosamplerwith Transfer Line
Catalog number MI-25118000
Price (USD)
-
Product Type:
Vial Loader and Tray Upgrade
Description:
Upgrade to 120 vial capacity
Powered by a completely new design, the Thermo Scientific™ TriPlus™ 500 Headspace Autosampler delivers reliability and robustness for testing laboratories performing volatiles analysis. Valve-and-loop technology and other innovative features ensure that this autosampler streamlines daily workflows, saving time with reliable unattended operation, optimizing sample throughput with expanded vial capability, and facilitating method portability with powerful software. Smartly integrated with the Thermo Scientific™ TRACE™ 1600 Series GC, this modular platform provides today's analytical testing and services laboratories with the productivity needed to succeed.
Reliability for analytical testing and services laboratories and versatility for research and method development.
The new design of the TriPlus 500 Headspace Autosampler incorporates innovative features for highly accurate analytical results, including:
New pneumatic controls for high precision and robustness
  • Best injection repeatability for consistent results at every run.
  • No carryover due to effective sample path purging.
  • Automatic leak check during injection to ensure safe runs.
Best sample integrity and ease-of-use via direct column connection
  • A shorter sample path for high inertness towards polar compounds.
  • Expanded high temperature applicability with higher recovery of high boiling point compounds.
  • Simplified method set up with no transfer line and injector temperature settings required.
 Compact yet scalable design for better laboratory efficiency and productivity
  • Compact HS-GC design saves 30% of valuable bench space.
  • Tailored throughput requirements with in-field scalability from 12 to 240 vials, requiring no additional space and no compromise on performance.
  • Faster sample equilibration with the new Quick Spin Shaking design for maximal sensitivity in a shorter cycle time.
 Comprehensive HS-GC configuration solution
  • Available with direct column connection or transfer line for maximum configuration flexibility.
  • When a direct column connection is not possible, the autosampler configured with a transfer line maintains high precision, efficient sample transfer.
  • When configured with a transfer line, the TriPlus 500 HS autosampler is ideal for  connecting to the Thermo Scientific™ TRACE™ 1600 Series GC with auxiliary oven or for connection to third-party GC instruments
  • The TriPlus 500 HS with transfer line allows combined headspace and liquid injection in the same column, through operation with the TriPlus 100 LS or TriPlus RSH or RSH SMART autosampler on the same GC. 
Software and system checks for compliance and data integrity
  • Integrated with Thermo Scientific™ Chromeleon™ CDS software, Title 21 CFR Part 11-compliant.
  • Easy analytical system performance  qualification with the IQ/OQ/PQ Qualification Wizard and daily sequence performance checks with the System Suitability Test Wizard.
Specifications
DescriptionUpgrade to 120 vial capacity
QuantityEach
SoftwareThermo Scientific Chromeleon CDS, Thermo Scientific TraceFinder, and Thermo Scientific XCalibur Software
For Use With (Equipment)TriPlus™ 500 GC Headspace Autosampler
Injection ModeVial Tray Only
Product TypeVial Loader and Tray Upgrade
Sample Capacity120 Vials
Unit SizeEach