Neon™ Transfection System 10 μL Kit, 96 x 2 Reactions - FAQs
Neon™ Transfection System 10 μL Kit, 96 x 2 Reactions - FAQs
View additional product information for Neon™ Transfection System 10 μL Kit - FAQs (MPK1096)
1 product FAQs found
Is the Neon Transfection System 10 μL-Kit (Cat. No. MPK1096, MPK1025) compatible with the Neon NxT Electroporation System (NEON1)?
No, the two systems are not compatible with each other. Neon tips won't fit into the Neon Nxt pipette. The tips, tubes and pipettes are all system specific.
Find additional tips, troubleshooting help, and resources within our RNAi Support Center.