MAGPIX™ System Automation Interface Module - FAQs
MAGPIX™ System Automation Interface Module - FAQs
View additional product information for MAGPIX™ System Automation Interface Module - FAQs (MPXAM)
2 product FAQs found
How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?
With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:
- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the MAGPIX System?
The MAGPIX System is a compact benchtop instrument with a multiplex capability of up to 50 analytes. It is a robust and cost-effective multiplexing tool. Streamlined startup and shutdown protocols, and minimal maintenance requirements, make the system easy to use-ideal for both new and experienced users. It features simple out-of-the-box setup and interactive software. The MAGPIX System analyzes magnetic beads immobilized with a magnet, excites the beads using light-emitting diodes (LEDs), and then detects and analyzes the beads using a CCD camera.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.