MAGPIX™ Calibration Kit
Invitrogen™

MAGPIX™ Calibration Kit

This convenient calibration kit contains xMAP™ calibrators for the MAGPIX™ system classification and reporter channels.
Have Questions?
Catalog NumberQuantity
MPXCALK2525 Reactions
Catalog number MPXCALK25
Price (USD)
663.65
Online exclusive
684.00
Save 20.35
Each
Quantity:
25 Reactions
Price (USD)
663.65
Online exclusive
684.00
Save 20.35
Each

This convenient calibration kit contains xMAP™ calibrators for the MAGPIX™ system classification and reporter channels. Calibration normalizes the settings for the system and helps ensure optimal and consistent microsphere classification. The kit is intended to be used with the off-plate reagent block. Each kit provides 25 calibrations.

Each Kit includes:

  • Stripwells
  • A lot-specific CD for importing target values

The MAGPIX™ system is a versatile, compact, fluorescent detection system based on Luminex's proven xMAP™ technology. The system is a simple immunoassay platform designed for the simultaneous analysis of up to 50 intracellular and extracellular markers in single biological samples. The MAGPIX™ system provides users with a powerful multiplexing tool that offers higher sample throughput, increased assay flexibility, reduced sample volume, and lower costs than traditional ELISA and Western blotting.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Product LineMAGPIX
Quantity25 Reactions
Shipping ConditionWet Ice
For Use With (Equipment)Flow Cytometer
Product TypeCalibration Kit
Unit SizeEach
Contents & Storage
• 25 strip wells (Cat. No. 13-52047)
• MAGPIX™ Calibrator Microspheres, 5 mL (MPXCAL-05)
• MAGPIX™ Calibration Kit CD (89-20233-00-001)

Store at 2°C to 8°C.

Frequently asked questions (FAQs)

How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?

With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:

- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.