Invitrogen™

MAGPIX™ Drive Fluid (4 pack)

Catalog number:  MPXDF4PK

 Related applications:

Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

MAGPIX® Drive Fluid is the delivery medium for transporting the sample being analyzed on the Luminex® MAGPIX® system to the optic components. Each unit in the 4-pack of drive fluid contains 700 mL.

Please refer to a Luminex® instrument user manual for proper installation and use of MAGPIX® Drive Fluid with the MAGPIX® System.

For research use only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Luminex® Instruments, MAGPIX® System
Product Line: MAGPIX®
Product Size: 4 x 700 mL
Shipping Condition: Room Temperature
Type: CatalogProductData

Contents & storage

4 × 700 mL

Documents

Manuals & protocols