Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader - FAQs
Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader - FAQs
View additional product information for Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader - FAQs (N16443, N03395, VLUPL0002, VLUPLAT08, N02697, VLUPD005, N02693, N02696, VLUPLA06, VLUPD209, N02339, SP-00094, SP-00096, 2805690, N02690, VLUPT004, N02692, N06210, F42550, N02691, VLUPA003, VLUPLT07, N03078, N03079, N16484, SP-00720, N16308S)
1 product FAQs found
Can the Varioskan LUX Multimode Microplate Reader be integrated with robotics?
Yes, it can. To do so, you must purchase the robotic tray separately (Cat. No. N03079).
The SkanIt Automation Interface manual is available from your local support team.