PRIMING VESSEL, MD COMBI NL NO - FAQs
PRIMING VESSEL, MD COMBI NL NO - FAQs
View additional product information for PRIMING VESSEL, MD COMBI NL NO - FAQs (N07271)
2 product FAQs found
Can the Multidrop Combi and Multidrop Combi nL priming vessel be autoclaved?
Yes, autoclaving conditions are as follows: 1 bar pressure at 121 degrees C for 20 minutes.
What is the material of the Multidrop Combi and Multidrop Combi nL priming vessel?
The Multidrop Combi and Multidrop Combi nL priming vessel is made of blue polypropylene (PP).